3-Fluoro-2-nitrotoluene 98%
Supplier: Apollo Scientific
Warning
PC2964-5G PC2964-1G PC2964-25G
APOSPC2964-5GEA 52.2 EUR
APOSPC2964-5G APOSPC2964-1G APOSPC2964-25G
3-Fluoro-2-nitrotoluene 98%
3-Fluoro-2-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Melting Pt: 17.5…18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08458093
CAS Number: 3013-27-2