1,2-Diiodotetrafluorobenzene 97%
Supplier: Apollo Scientific
Warning
PC2969-1G PC2969-25G PC2969-5G
APOSPC2969-1GEA 36.4 EUR
APOSPC2969-1G APOSPC2969-25G APOSPC2969-5G
1,2-Diiodotetrafluorobenzene 97%
1,2-Diiodotetrafluorobenzene
Formula: C₆F₄I₂
MW: 401.87 g/mol
Melting Pt: 49…50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00019011
CAS Number: 2708-97-6