1,4-Diiodotetrafluorobenzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1,2,4,5-Tetrafluoro-3,6-diiodobenzene
PC2970-1G PC2970-25G
APOSPC2970-1GEA 29 EUR
APOSPC2970-1G APOSPC2970-25G
1,4-Diiodotetrafluorobenzene 97%
1,4-Diiodotetrafluorobenzene
Formula: C₆F₄I₂
MW: 401.87 g/mol
Melting Pt: 107…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001055
CAS Number: 392-57-4
EINECS: 206-877-8