3-Bromo-5-chloro-2-fluoropyridine
Supplier: Apollo Scientific
Warning
PC2973-25G PC2973-5G
APOSPC2973-25GEA 59.6 EUR
APOSPC2973-25G APOSPC2973-5G
3-Bromo-5-chloro-2-fluoropyridine
3-Bromo-5-chloro-2-fluoropyridine
Formula: C₅H₂BrClFN
Melting Pt: 55.6…55.8 °C
Storage Temperature: Ambient
MDL Number: MFCD04972409
CAS Number: 884494-87-5