2-(Trifluoromethyl)benzenesulphonamide
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
α,α,α-Trifluoro-o-toluenesulphonamide
PC2977-1G
PC2977-25G
PC2977-5G
APOSPC2977-1GEA
21.9
EUR
APOSPC2977-1G
APOSPC2977-25G
APOSPC2977-5G
2-(Trifluoromethyl)benzenesulphonamide
2-(Trifluoromethyl)benzenesulphonamide
Formula:
C₇H₆F₃NO₂S MW: 225.19 Melting Pt: 180…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042420 CAS Number: 1869-24-5 |