1,1,2,2-Tetrafluoro-1,2-diiodoethane 97%
Supplier: Apollo Scientific
Warning
PC2980-25G PC2980-5G
APOSPC2980-25GEA 499 EUR
APOSPC2980-25G APOSPC2980-5G
1,1,2,2-Tetrafluoro-1,2-diiodoethane 97%
1,1,2,2-Tetrafluoro-1,2-diiodoethane
Formula: C₂F₄I₂
Boiling Pt: 112…113 °C
Melting Pt: –21 °C
Density: 2.629 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00039401
CAS Number: 354-65-4
EINECS: 205-567-2