3-Amino-6-fluoro-2-methylpyridine
Supplier: Apollo Scientific
Warning
Synonyms: 6-Fluoro-2-methylpyridin-3-amine
PC2988-1G PC2988-5G PC2988-25G
APOSPC2988-1GEA 79.9 EUR
APOSPC2988-1G APOSPC2988-5G APOSPC2988-25G
3-Amino-6-fluoro-2-methylpyridine
3-Amino-6-fluoro-2-methylpyridine
Formula: C₆H₇FN₂
MW: 126.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094949
CAS Number: 28489-47-6