2,2-Difluoroethyl-p-toluenesulphonate 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2-Difluoroethyl 4-methylbenzenesulphonate
PC3000-5G PC3000-1G
APOSPC3000-5GEA 143 EUR
APOSPC3000-5G APOSPC3000-1G
2,2-Difluoroethyl-p-toluenesulphonate 97%
2,2-Difluoroethyl-p-toluenesulphonate
Formula: C₉H₁₀F₂O₃S
MW: 236.24 g/mol
Melting Pt: 30…31 °C
Storage Temperature: Ambient
MDL Number: MFCD04038284
CAS Number: 135206-84-7