1-(3-Fluoro-4-methylphenyl)-5-oxopyrrolidine-3-carboxylic acid
Supplier: Apollo Scientific
PC300002-1G PC300002-5G PC300002-10G
APOSPC300002-1GEA 163 EUR
APOSPC300002-1G APOSPC300002-5G APOSPC300002-10G
1-(3-Fluoro-4-methylphenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(3-Fluoro-4-methylphenyl)-5-oxopyrrolidine-3-carboxylic acid
Formula: C₁₂H₁₂FNO₃
MW: 237.23 g/mol
Melting Pt: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD07359954
CAS Number: 63674-46-4