N-(tert-Butoxycarbonyl)-3,4-difluoro-D-phenylalanine 98%
Supplier: Apollo Scientific
Synonyms: Boc-D-Phe(3,4-F)-OH, N-Boc-3,4-difluoro-D-phenylalanine, Boc-D-Phe(3,4-DiF)-OH
PC3004-25G PC3004-5G
APOSPC3004-25GEA 583 EUR
APOSPC3004-25G APOSPC3004-5G
N-(tert-Butoxycarbonyl)-3,4-difluoro-D-phenylalanine 98%
N-(tert-Butoxycarbonyl)-3,4-difluoro-D-phenylalanine
Formula: C₁₄H₁₇F₂NO₄
Storage Temperature: Ambient
MDL Number: MFCD00672519
CAS Number: 205445-51-8