2-Hydroxy-5-(trifluoromethyl)aniline 95%
Supplier: Apollo Scientific
Warning
PC300506-1G PC300506-250MG PC300506-5G
APOSPC300506-1GEA 58 EUR
APOSPC300506-1G APOSPC300506-250MG APOSPC300506-5G
2-Hydroxy-5-(trifluoromethyl)aniline 95%
2-Hydroxy-5-(trifluoromethyl)aniline
Formula: C₇H₆F₃NO
Melting Pt: 118…120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01731750
CAS Number: 454-81-9