4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
Supplier: Apollo Scientific
Warning
PC300540-1G PC300540-250MG
APOSPC300540-1GEA 183 EUR
APOSPC300540-1G APOSPC300540-250MG
4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
Formula: C₁₃H₇ClF₃NO₃
Melting Pt: 156…159 °C
Storage Temperature: Ambient
MDL Number: MFCD11840267
CAS Number: 105626-87-7