2-Methyl-6-(trifluoromethyl)pyridine-3-carboxaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 2-Methyl-6-(trifluoromethyl)nicotinaldehyde
PC300636-1G PC300636-250MG
APOSPC300636-1GEA 186 EUR
APOSPC300636-1G APOSPC300636-250MG
2-Methyl-6-(trifluoromethyl)pyridine-3-carboxaldehyde
2-Methyl-6-(trifluoromethyl)pyridine-3-carboxaldehyde
Formula: C₈H₆F₃NO
MW: 189.137 g/mol
Melting Pt: 45…47 °C
Storage Temperature: Ambient
MDL Number: MFCD11035943
CAS Number: 545394-83-0