Ethyl-2-amino-4-(4-fluorophenyl)thiophene-3-carboxylate
Supplier: Apollo Scientific
PC300687-10G PC300687-1G PC300687-5G
APOSPC300687-10GEA 302 EUR
APOSPC300687-10G APOSPC300687-1G APOSPC300687-5G
Ethyl-2-amino-4-(4-fluorophenyl)thiophene-3-carboxylate
Ethyl-2-amino-4-(4-fluorophenyl)thiophene-3-carboxylate
Formula: C₁₃H₁₂FNO₂S
MW: 265.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00239291
CAS Number: 35978-33-7