3-(Trifluoromethoxy)benzamidoxime
Supplier: Apollo Scientific
|
Warning
|
PC300708-1G
PC300708-5G
PC300708-25G
APOSPC300708-1GEA
21.9
EUR
APOSPC300708-1G
APOSPC300708-5G
APOSPC300708-25G
3-(Trifluoromethoxy)benzamidoxime
3-(Trifluoromethoxy)benzamidoxime
Formula:
C₈H₇F₃N₂O₂ Melting Pt: 50…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD06660166 CAS Number: 886500-80-7 |