4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
PC300799-1G PC300799-5G
APOSPC300799-1GEA 109 EUR
APOSPC300799-1G APOSPC300799-5G
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonyl chloride
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonyl chloride
Formula: C₁₂H₇ClF₃NO₃S
MW: 337.697 g/mol
Melting Pt: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD07774164
CAS Number: 874839-13-1
UN: 3261
ADR: 8 III