(4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}phenyl)methanol
Supplier: Apollo Scientific
Synonyms: (4-((6-(Trifluoromethyl)pyridin-2-yl)oxy)phenyl)methanol
PC300941-1G PC300941-5G
APOSPC300941-1GEA 249 EUR
APOSPC300941-1G APOSPC300941-5G
(4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}phenyl)methanol
(4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}phenyl)methanol
Formula: C₁₃H₁₀F₃NO₂
MW: 269.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10703547
CAS Number: 1086376-61-5