3,5-Difluorobenzamidoxime 97%
Supplier: Apollo Scientific
PC302038-10G PC302038-1G PC302038-5G
APOSPC302038-10GEA 79.9 EUR
APOSPC302038-10G APOSPC302038-1G APOSPC302038-5G
3,5-Difluorobenzamidoxime 97%
3,5-Difluorobenzamidoxime
Formula: C₇H₆F₂N₂O
Storage Temperature: Ambient
CAS Number: 874880-59-8