2-(3-Chloro-5-(trifluoromethyl)phenyl)acetonitrile 97%
Supplier: Apollo Scientific
PC302052-1G PC302052-25G
APOSPC302052-1GEA 21.9 EUR
APOSPC302052-1G APOSPC302052-25G
2-(3-Chloro-5-(trifluoromethyl)phenyl)acetonitrile 97%
2-(3-Chloro-5-(trifluoromethyl)phenyl)acetonitrile
Formula: C₉H₅ClF₃N
MW: 219.5909 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115850
CAS Number: 886496-95-3