3-Fluoro-4-(trifluoromethoxy)benzylamine 98%
Supplier: Apollo Scientific
PC302053-1G PC302053-5G
APOSPC302053-1GEA 134 EUR
APOSPC302053-1G APOSPC302053-5G
3-Fluoro-4-(trifluoromethoxy)benzylamine 98%
3-Fluoro-4-(trifluoromethoxy)benzylamine
Formula: C₈H₇F₄NO
MW: 209.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660207
CAS Number: 886499-13-4