(4-Chloro-3-(trifluoromethyl)phenyl)acetic acid 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloro-3-(trifluoromethyl)phenylacetic acid
PC302061-1G PC302061-5G PC302061-10G PC302061-25G
APOSPC302061-1GEA 33.5 EUR
APOSPC302061-1G APOSPC302061-5G APOSPC302061-10G APOSPC302061-25G
(4-Chloro-3-(trifluoromethyl)phenyl)acetic acid 97%
(4-Chloro-3-(trifluoromethyl)phenyl)acetic acid
Formula: C₉H₆ClF₃O₂
MW: 238.59 g/mol
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD04116012
CAS Number: 22902-86-9