2-Chloro-3,6-difluorobenzonitrile 98%
Supplier: Apollo Scientific
Warning
PC302072-25G PC302072-5G PC302072-1G
APOSPC302072-25GEA 264 EUR
APOSPC302072-25G APOSPC302072-5G APOSPC302072-1G
2-Chloro-3,6-difluorobenzonitrile 98%
2-Chloro-3,6-difluorobenzonitrile
Formula: C₇H₂ClF₂N
Storage Temperature: Ambient
MDL Number: MFCD06660175
CAS Number: 886501-33-3
UN: 3276
ADR: 6.1,III