2-Methyl-5-(trifluoromethoxy)benzylamine 98%
Supplier: Apollo Scientific
PC302081-1G PC302081-5G
APOSPC302081-1GEA 116 EUR
APOSPC302081-1G APOSPC302081-5G
2-Methyl-5-(trifluoromethoxy)benzylamine 98%
2-Methyl-5-(trifluoromethoxy)benzylamine
Formula: C₉H₁₀F₃NO
MW: 205.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201056
CAS Number: 1373920-87-6