3,5-Difluoro-2-methoxybenzylamine 98%
Supplier: Apollo Scientific
PC302084-1G PC302084-5G PC302084-10G
APOSPC302084-1GEA 156 EUR
APOSPC302084-1G APOSPC302084-5G APOSPC302084-10G
3,5-Difluoro-2-methoxybenzylamine 98%
3,5-Difluoro-2-methoxybenzylamine
Formula: C₈H₉F₂NO
MW: 173.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660178
CAS Number: 886502-00-7