3-Methoxy-2-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
PC302094-1G PC302094-5G PC302094-10G
APOSPC302094-1GEA 101 EUR
APOSPC302094-1G APOSPC302094-5G APOSPC302094-10G
3-Methoxy-2-(trifluoromethyl)benzaldehyde 98%
3-Methoxy-2-(trifluoromethyl)benzaldehyde
Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185677
CAS Number: 1214384-89-0