2-Bromo-6-chloro-4-fluorophenol 97%
Supplier: Apollo Scientific
Danger
PC302166-5G PC302166-1G
APOSPC302166-5GEA 65.4 EUR
APOSPC302166-5G APOSPC302166-1G
2-Bromo-6-chloro-4-fluorophenol 97%
2-Bromo-6-chloro-4-fluorophenol
Formula: C₆H₃BrClFO
MW: 225.44 g/mol
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD04115954
CAS Number: 886499-83-8