2-Methoxy-4-(trifluoromethoxy)benzoic acid 98%
Supplier: Apollo Scientific
Warning
PC302174-10G PC302174-5G PC302174-1G
APOSPC302174-10GEA 266 EUR
APOSPC302174-10G APOSPC302174-5G APOSPC302174-1G
2-Methoxy-4-(trifluoromethoxy)benzoic acid 98%
2-Methoxy-4-(trifluoromethoxy)benzoic acid
Formula: C₉H₇F₃O₄
MW: 236.15 g/mol
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD04115966
CAS Number: 848947-91-1