3-Chloro-2-fluoro-6-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
PC302193-1G PC302193-5G
APOSPC302193-1GEA 261 EUR
APOSPC302193-1G APOSPC302193-5G
3-Chloro-2-fluoro-6-(trifluoromethyl)aniline 98%
3-Chloro-2-fluoro-6-(trifluoromethyl)aniline
Formula: C₇H₄ClF₄N
MW: 213.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519381
CAS Number: 1092461-38-5
UN: 2810
ADR: 6.1 III