2-Methoxy-4,6-bis(trifluoromethyl)benzonitrile 98%
Supplier: Apollo Scientific
PC302254-1G PC302254-5G PC302254-10G PC302254-25G PC302254-100G
APOSPC302254-1GEA 102 EUR
APOSPC302254-1G APOSPC302254-5G APOSPC302254-10G APOSPC302254-25G APOSPC302254-100G
2-Methoxy-4,6-bis(trifluoromethyl)benzonitrile 98%
2-Methoxy-4,6-bis(trifluoromethyl)benzonitrile
Formula: C₁₀H₅F₆NO
MW: 269.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519321
CAS Number: 1092460-75-7