3-Ethoxy-2,6-difluorobenzoic acid 97%
Supplier: Apollo Scientific
Warning
PC302296-5G PC302296-10G PC302296-1G
APOSPC302296-5GEA 84.1 EUR
APOSPC302296-5G APOSPC302296-10G APOSPC302296-1G
3-Ethoxy-2,6-difluorobenzoic acid 97%
3-Ethoxy-2,6-difluorobenzoic acid
Formula: C₉H₈F₂O₃
MW: 202.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519368
CAS Number: 651734-62-2