3'-Fluoro-5'-(trifluoromethoxy)acetophenone 97%
Supplier: Apollo Scientific
Warning
PC302302-1G PC302302-5G
APOSPC302302-1GEA 224 EUR
APOSPC302302-1G APOSPC302302-5G
3'-Fluoro-5'-(trifluoromethoxy)acetophenone 97%
3'-Fluoro-5'-(trifluoromethoxy)acetophenone
Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18783230
CAS Number: 1352999-53-1