3-Methoxy-4-(trifluoromethyl)benzamide
Supplier: Apollo Scientific
PC302306-1G PC302306-5G
APOSPC302306-1GEA 159 EUR
APOSPC302306-1G APOSPC302306-5G
3-Methoxy-4-(trifluoromethyl)benzamide
3-Methoxy-4-(trifluoromethyl)benzamide
Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185679
CAS Number: 1214328-36-5