4-Ethoxy-3-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
PC302332-5G PC302332-10G PC302332-1G
APOSPC302332-5GEA 43.5 EUR
APOSPC302332-5G APOSPC302332-10G APOSPC302332-1G
4-Ethoxy-3-(trifluoromethyl)aniline 98%
4-Ethoxy-3-(trifluoromethyl)aniline
Formula: C₉H₁₀F₃NO
MW: 205.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08056137
CAS Number: 2713-74-8