4-Methoxy-3-(trifluoromethoxy)benzyl alcohol
Supplier: Apollo Scientific
PC302346-1G PC302346-5G
APOSPC302346-1GEA 180 EUR
APOSPC302346-1G APOSPC302346-5G
4-Methoxy-3-(trifluoromethoxy)benzyl alcohol
4-Methoxy-3-(trifluoromethoxy)benzyl alcohol
Formula: C₉H₉F₃O₃
MW: 221.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18398472
CAS Number: 853771-91-2