6-Chloro-2-fluoro-3-methoxybenzaldehyde 97%
Supplier: Apollo Scientific
Warning
PC302367-5G PC302367-10G PC302367-1G
APOSPC302367-5GEA 122 EUR
APOSPC302367-5G APOSPC302367-10G APOSPC302367-1G
6-Chloro-2-fluoro-3-methoxybenzaldehyde 97%
6-Chloro-2-fluoro-3-methoxybenzaldehyde
Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
Melting Pt: 95…100 °C
Storage Temperature: Ambient
MDL Number: MFCD04115946
CAS Number: 112641-64-2