4-n-Butoxy-2,6-difluorophenol 98%
Supplier: Apollo Scientific
Synonyms: 4-Butoxy-2,6-difluorophenol
PC302406-1G PC302406-5G PC302406-10G
APOSPC302406-1GEA 148 EUR
APOSPC302406-1G APOSPC302406-5G APOSPC302406-10G
4-n-Butoxy-2,6-difluorophenol 98%
4-n-Butoxy-2,6-difluorophenol
Formula: C₁₀H₁₂F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22201025
CAS Number: 1373920-65-0