2,3-Difluoro-4-methoxy-DL-phenylalanine
Supplier: Apollo Scientific
PC302431-1G
PC302431-5G
APOSPC302431-1GEA
207
EUR
APOSPC302431-1G
APOSPC302431-5G
2,3-Difluoro-4-methoxy-DL-phenylalanine
2,3-Difluoro-4-methoxy-DL-phenylalanine
Formula:
C₁₀H₁₁F₂NO₃ MW: 231.20 g/mol Melting Pt: 260 °C Storage Temperature: Ambient |
MDL Number:
MFCD09832370 CAS Number: 1256482-63-9 |