2,4-Difluoro-3-methoxybenzamide
Supplier: Apollo Scientific
PC302437-10G PC302437-5G
APOSPC302437-10GEA 282 EUR
APOSPC302437-10G APOSPC302437-5G
2,4-Difluoro-3-methoxybenzamide
2,4-Difluoro-3-methoxybenzamide
Formula: C₈H₇F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD04115931
CAS Number: 479091-06-0