2,4-Difluoro-3-methoxybenzoic acid 98%
Supplier: Apollo Scientific
Warning
PC302438-25G PC302438-5G PC302438-1G
APOSPC302438-25GEA 366 EUR
APOSPC302438-25G APOSPC302438-5G APOSPC302438-1G
2,4-Difluoro-3-methoxybenzoic acid 98%
2,4-Difluoro-3-methoxybenzoic acid
Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
Melting Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD02258905
CAS Number: 178974-97-5