2-Fluoro-4-(pentafluorothio)benzyl bromide
Supplier: Apollo Scientific
Danger
PC302452-1G PC302452-5G
APOSPC302452-1GEA 290 EUR
APOSPC302452-1G APOSPC302452-5G
2-Fluoro-4-(pentafluorothio)benzyl bromide
2-Fluoro-4-(pentafluorothio)benzyl bromide
Formula: C₇H₅BrF₆S
MW: 315.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652507
CAS Number: 1240257-17-3
UN: 3265
ADR: 8,III