3-(4-Ethoxy-3,5-difluorophenyl)propionic acid 98%
Supplier: Apollo Scientific
PC302488-1G PC302488-5G
APOSPC302488-1GEA 116 EUR
APOSPC302488-1G APOSPC302488-5G
3-(4-Ethoxy-3,5-difluorophenyl)propionic acid 98%
3-(4-Ethoxy-3,5-difluorophenyl)propionic acid
Formula: C₁₁H₁₂F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD11519343
CAS Number: 1092461-00-1