3-(2-Fluoro-4-(trifluoromethoxy)phenyl)propionic acid 98%
Supplier: Apollo Scientific
PC302499-1G PC302499-5G PC302499-10G
APOSPC302499-1GEA 109 EUR
APOSPC302499-1G APOSPC302499-5G APOSPC302499-10G
3-(2-Fluoro-4-(trifluoromethoxy)phenyl)propionic acid 98%
3-(2-Fluoro-4-(trifluoromethoxy)phenyl)propionic acid
Formula: C₁₀H₈F₄O₃
MW: 252.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652497
CAS Number: 1240257-16-2