3-(4-(Difluoromethoxy)-3-methoxyphenyl)propionic acid 97%
Supplier: Apollo Scientific
PC302512-1G PC302512-5G
APOSPC302512-1GEA 116 EUR
APOSPC302512-1G APOSPC302512-5G
3-(4-(Difluoromethoxy)-3-methoxyphenyl)propionic acid 97%
3-(4-(Difluoromethoxy)-3-methoxyphenyl)propionic acid
Formula: C₁₁H₁₂F₂O₄
Melting Pt: 66…69 °C
Storage Temperature: Ambient
MDL Number: MFCD11519324
CAS Number: 1092460-76-8