3'-Chloro-5'-(trifluoromethyl)acetophenone
Supplier: Apollo Scientific
Warning
PC302531-25G PC302531-5G
APOSPC302531-25GEA 449 EUR
APOSPC302531-25G APOSPC302531-5G
3'-Chloro-5'-(trifluoromethyl)acetophenone
3'-Chloro-5'-(trifluoromethyl)acetophenone
Formula: C₉H₆ClF₃O
MW: 222.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115856
CAS Number: 886497-11-6