3'-Chloro-5'-fluoro-4'-methoxyacetophenone 97%
Supplier: Apollo Scientific
PC302535-10G PC302535-5G PC302535-1G
APOSPC302535-10GEA 360 EUR
APOSPC302535-10G APOSPC302535-5G APOSPC302535-1G
3'-Chloro-5'-fluoro-4'-methoxyacetophenone 97%
3'-Chloro-5'-fluoro-4'-methoxyacetophenone
Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD04115862
CAS Number: 886497-19-4