4-Difluoromethoxy-2-fluorobenzoic acid 98%
Supplier: Apollo Scientific
PC302560-1G PC302560-5G
APOSPC302560-1GEA 226 EUR
APOSPC302560-1G APOSPC302560-5G
4-Difluoromethoxy-2-fluorobenzoic acid 98%
4-Difluoromethoxy-2-fluorobenzoic acid
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Melting Pt: 135…138 °C
Storage Temperature: Ambient
MDL Number: MFCD09832367
CAS Number: 1214347-60-0