4-Chloro-3-ethoxy-2-fluorobenzyl bromide 98%
Supplier: Apollo Scientific
Synonyms: 4-Chloro-3-ethoxy-2-fluorobenzyl bromide
PC302579-1G PC302579-5G PC302579-10G
APOSPC302579-1GEA 116 EUR
APOSPC302579-1G APOSPC302579-5G APOSPC302579-10G
4-Chloro-3-ethoxy-2-fluorobenzyl bromide 98%
4-Chloro-3-ethoxy-2-fluorobenzyl bromide
Formula: C₉H₉BrClFO
MW: 267.52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687186
CAS Number: 1323966-30-8