4-Methoxy-2-(trifluoromethyl)benzoyl chloride 97%
Supplier: Apollo Scientific
Danger
PC302600-5G PC302600-10G PC302600-1G
APOSPC302600-5GEA 116 EUR
APOSPC302600-5G APOSPC302600-10G APOSPC302600-1G
4-Methoxy-2-(trifluoromethyl)benzoyl chloride 97%
4-Methoxy-2-(trifluoromethyl)benzoyl chloride
Formula: C₉H₆ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09025403
CAS Number: 98187-17-8
UN: 3265
ADR: 8,II