5-(Difluoromethoxy)-2-fluorobenzyl bromide
Supplier: Apollo Scientific
Danger
PC302610-1G PC302610-5G
APOSPC302610-1GEA 259 EUR
APOSPC302610-1G APOSPC302610-5G
5-(Difluoromethoxy)-2-fluorobenzyl bromide
5-(Difluoromethoxy)-2-fluorobenzyl bromide
Formula: C₈H₆BrF₃O
MW: 255.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832361
CAS Number: 1017779-36-0
UN: 3265
ADR: 8,III