5-Fluoro-2-(trifluoromethoxy)benzyl bromide 98%
Supplier: Apollo Scientific
PC302619-1G
PC302619-5G
PC302619-10G
APOSPC302619-1GEA
116
EUR
APOSPC302619-1G
APOSPC302619-5G
APOSPC302619-10G
5-Fluoro-2-(trifluoromethoxy)benzyl bromide 98%
5-Fluoro-2-(trifluoromethoxy)benzyl bromide
Formula:
C₈H₅BrF₄O MW: 273.02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11519337 CAS Number: 1092460-88-2 UN: 3265 ADR: 8 III |